Learn More
R&D Systems™ Recombinant Human VSIG8 Fc Chimera Biotinylated Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Sodium Acetate and Mannitol with Trehalose. |
| Gene ID (Entrez) | 391123 |
| Molecular Weight (g/mol) | M.W. (Observed): 56-67 kDa, reducing conditions; M.W. (theoretical): 54 kDa (unlabeled) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human VSIG8 protein Human VSIG8 (Val22-Gly263) (N-terminus) Accession # Q5VU13 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Biotin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.