Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VSTM4 Fc Chimera Protein

Catalog No. 2086VT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-86V-T050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 20-86V-T050 Supplier R&D Systems™ Supplier No. 2086VT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3 antibody induced IFN-gamma secretion by human peripheral blood mononuclear cells (PBMC). The ED50 for this effect is 1-10 μg/mL.

Specifications

Accession Number Q8IW00.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 196740
Molecular Weight (g/mol) M.W. (Observed): 58-74 kDa, reducing conditions; M.W. (theoretical): 45 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human VSTM4 protein Human VSTM4 (Leu24-Tyr180) (N-terminus) Accession # Q8IW00.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, ≤ -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, ≤ -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity VSTM4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VSTM4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.