Learn More
R&D Systems™ Recombinant Human Wnt-10b Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
| Gene ID (Entrez) | 7480 |
| Molecular Weight (g/mol) | 40 kDa |
| Name | Wnt-10b |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Wnt-10b protein Met1-Lys389 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, Wnt10b, WNT12, WNT-12 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.