Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Wnt/RSPO1 Agonist Protein, CF
SDP

Catalog No. BTWRSP1500 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTWRSP1500 500 μg
BTWRSP1100 100 μg
BTWRSP101M 1 mg
BTWRSP1050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. BTWRSP1500 Supplier R&D Systems™ Supplier No. BTWRSP1500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to activate TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 7.00-84.0 ng/mL.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Molecular Weight (g/mol) 86-96 kDa and >190 kDa, under reducing conditions.
Quantity 500 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Wnt/RSPO1 protein Proprietary
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CRISTIN3, roof plate-specific spondin-1, RSPO, RSPO 1, R-spondin homolog, R-spondin-1, Wnt
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500μg/mL in PBS.
Form Lyophilized
Protein Wnt/RSPO1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.