Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human XCL1/Lymphotactin Protein
SDP

Catalog No. 695LT025CF Shop All R&D Systems Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
695LT025CF Lyophilized from a 0.2 μm filtered solution in PBS.
695LT025 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 695LT025CF Supplier R&D Systems™ Supplier No. 695LT025/CF
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human XCR1. The ED50 for this effect is 0.0800-0.500 μg/mL.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 6375
Molecular Weight (g/mol) 10.4 kDa
Name XCL1/Lymphotactin
Quantity 25 μg
Source E. coli-derived human XCL1/Lymphotactin protein Val22-Gly114,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.