Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 331 |
| Molecular Weight (g/mol) | Observed Molecular Wt: 54-56kDa, reducing conditions, Predicted Molecular Wt.: 58kDa |
| Name | XIAP Full Length |
| Quantity | 50 μg |
| Source | E. coli-derived human XIAP protein, (N-terminus) ATVID 6-His tag SSNG Human XIAP (Met1-Ser497) Accession &Num; AAC50373 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 100 μg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
