Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Xylulokinase/XYLB Protein

Catalog No. 8267XB020
Change view
Click to view available options
20ug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
82-67X-B020 20ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 82-67X-B020 Supplier R&D Systems™ Supplier No. 8267XB020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to phosphorylate D-Xylulose. The specific activity is >6,000 pmol/min/μg, as measured under the described conditions.

Order Info

The product is shipped with dry ice or equivalent. Upon receipt, store it immediately

Specifications

Accession Number O75191
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT and Glycerol.
Gene ID (Entrez) 9942
Molecular Weight (g/mol) M.W. (Observed): 56 kDa, reducing conditions; M.W. (theoretical): 59 kDa
Name Xylulokinase/XYLB, Carrier Free
Quantity 20 μg
Source E. coli-derived human Xylulokinase/XYLB protein Ala2-Glu536, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol XYLB
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein Xylulokinase/XYLB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.