Learn More
Novus Biologicals™ Recombinant Influenza A H5N1 Hemagglutinin (A/Vietnam/HN31242/2007) His Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol |
| Molecular Weight (g/mol) | 37.4kDa |
| Name | Influenza A Haemagglutinin H5 Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Purity or Quality Grade | >90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.