Learn More
Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Lyophilized from a 0.22 μm filtered solution in Tris buffer (100 mM Tris, 150 mM NaCl, pH 8), preservative- and carrier-free. |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 40.44 kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -20°C to -80°C. Avoid freeze/thaw cycles. |
| Conjugate | Unconjugated |
| Species | Insect |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute with sterile deionized water with or without azide. |
| Form | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
