Learn More
Description
Specifications
Specifications
| Accession Number | K9N638 |
| For Use With (Application) | Bioactivity |
| Formulation | X mg/ml (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT |
| Molecular Weight (g/mol) | M.W.-theoretical: 37 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived mers-cov Papain-like Protease protein Leu1541 - Ser1859 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 100 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
| Protein | Papain-like Protease |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
