Learn More
Active Motif Recombinant METTL16 protein
SDP
Supplier:  Active Motif 81085
Encompass_Preferred
Recombinant human METTL16 protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_076991.3) with an N-terminal FLAG tag. The molecular weight of the protein is 64.9 kDa.
Catalog No. NC1851446
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.