Learn More
Apexbio Technology LLC Recombinant Mouse Prolactin 500ug
SDP
Supplier:  Apexbio Technology LLC P12260.5
Encompass_Preferred
Recombinant Mouse Prolactin (Uniprot P06879 full length tag-free) expressed in E coli is a peptide hormone belonging to the growth hormone/prolactin protein family Mouse prolactin consists of 197 amino acids and demonstrates structural similarity with human and rat counterparts sharing approximately 77%-90% amino acid sequence identity Prolactin primarily functions through interaction with cell-surface prolactin receptors to activate downstream signaling cascades including JAK-STAT pathways While traditionally associated with lactogenesis prolactin regulates diverse physiological processes in vertebrates including reproductive functions immune modulation and cellular growth Recombinant mouse prolactin serves as a valuable experimental tool for exploring hormone-receptor interactions signal transduction mechanisms and functional roles within endocrine and reproductive biology research
Catalog No. 50-000-63130
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.