Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse 4-1BB/TNFRSF9 Fc Chimera Protein

Catalog No. 9374B050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9374B050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9374B050 Supplier R&D Systems™ Supplier No. 9374B050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured by its ability to block 4-1BBL-induced IL-2 secretion by mouse T cells. The ED50 for this effect is 0.3-1 μg/mL in the presence of 0.4 μg/mL of recombinant mouse 4-1BBL. Measured by its ability to bind recombinant mouse 4-1BB Ligand in a functional ELISA.

Specifications

Accession Number P20334
For Use With (Application) Bioactivity, Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3604
Molecular Weight (g/mol) M.W. (Observed): 62-66 kDa, reducing conditions; M.W. (theoretical): 44.3 kDa (monomer)
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse 4-1 BB/TNFRSF9/CD137 protein Mouse 4-1 BB (Val24-Leu187) (N-terminus) Accession # P20334 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tnfrsf9
Product Type 4-1BB/TNFRSF9/CD137 Fc Chimera
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein 4-1 BB/TNFRSF9/CD137
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.