Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse ACBP Isoform 2 Protein
SDP

Catalog No. NBP26199605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26199605 0.5 mg
NB261996100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26199605 Supplier Novus Biologicals™ Supplier No. NBP2619960.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-87 of Mouse ACBP The Recombinant Mouse ACBP Isoform 2 Protein is derived from E. coli. The Recombinant Mouse ACBP Isoform 2 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate buffer saline (pH7.4)
Gene ID (Entrez) 1622
Molecular Weight (g/mol) 12.4 kDa
Quantity 0.5 mg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol DBI
Cross Reactivity ACBP
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.