Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Active Heparanase/HPSE Protein

Catalog No. 9788GH005
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
97-88G-H005 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 97-88G-H005 Supplier R&D Systems™ Supplier No. 9788GH005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to release biotinylated heparan sulfate from Recombinant Human Syndecan-4 (Catalog # 2918-SD ). 20 ng rmHPSE digestion will result in >50% of OD reduction compared with the Negative Control.

Specifications

Accession Number Q6YGZ1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and E64.
Gene ID (Entrez) 10855
Molecular Weight (g/mol) M.W. (Observed): 48-56 kDa (50 kDa subunit) and 8-12 kDa (8 kDa subunit), reducing conditions; M.W. (theoretical): 43 kDa (50 kDa subunit) and 9 kDa (8 kDa subunit)
Quantity 5 μg
Source Mouse myeloma cell line, NS0-derived mouse Heparanase/HPSE protein Asp28-Ile535, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity Heparanase/HPSE
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Heparanase/HPSE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.