Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Activin B Protein

Catalog No. 8260AB010CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
82-60A-B010CF Lyophilized from a 0.2μm filtered solution in HCl.
82-60A-B010 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 82-60A-B010CF Supplier R&D Systems™ Supplier No. 8260AB010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Activin B Protein is derived from HEK293. The Recombinant Mouse Activin B Protein has been validated for the following applications: Binding Activity.

Order Info

The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.

Specifications

Accession Number Q04999
Concentration Reconstitute at 100μg/mL in sterile 4mM HCl.
Formulation Lyophilized from a 0.2μm filtered solution in HCl.
Molecular Weight (g/mol) 13 kDa (monomer)
Name Activin B, Carrier Free
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived mouse Activin B protein Gly297-Ala411
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.