Learn More
R&D Systems™ Recombinant Mouse Activin RIIB Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 40.5 kDa (monomer) |
| Name | Activin RIIB Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Activin RIIB protein, (N-terminus) Mouse Activin RIIB (Ser19-Thr134) Accession &Num; P27040 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActivinRIIB, ACTRIIB, ACTR-IIB, ACVR2B, EC 2.7.11, EC 2.7.11.30, MGC116908 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.