Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Angiopoietin-like 3 (aa 17-220) Biotin

Catalog No. BT8344050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT8344050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT8344050 Supplier R&D Systems™ Supplier No. BT8344050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Angiopoietin-like 3 (aa 17-220) Biotin is derived from CHO. The Recombinant Mouse Angiopoietin-like 3 (aa 17-220) Biotin has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9R182
For Use With (Application) Bioactivity
Gene ID (Entrez) 27329
Molecular Weight (g/mol) Theoretical M.W.: 23kDa (unlabeled)
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Angiopoietin-like Protein 3/ANGPTL3 protein Ser17-Thr206,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2
Common Name Angiopoietin-like Protein 3/ANGPTL3
Gene Symbol Angptl3
Biological Activity Measured by its ability to promote the expansion of E16 rat liver mononuclear cells in vitro, in the presence of Recombinant Mouse SCF/c-kit Ligand, Recombinant Mouse Thrombopoietin/Tpo, and Recombinant Mouse Flt-3 Ligan. The ED50 for this effect is typically 20-120ng/mL in the presence of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody.
Conjugate Biotin
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser17-Thr206, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.