Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Angiopoietin-like 3 (HA-Tag) Protein

Catalog No. 9899AN050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-99AN-050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-99AN-050 Supplier R&D Systems™ Supplier No. 9899AN050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Angiopoietin-like 3 (HA-Tag) Protein is derived from NS0. The Recombinant Mouse Angiopoietin-like 3 (HA-Tag) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9R182
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and CHAPS with Trehalose.
Gene ID (Entrez) 27329
Molecular Weight (g/mol) 52 kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Angiopoietin-like protein, (N-terminus) 3/ANGPTL3 protein, (N-terminus) Hemagglutinin Tag (YPYDVPDYA) Mouse Angiopoietin-like protein, (N-terminus) 3 (Ser17-Thr455) Accession &Num; Q9R182 (C-terminus)
Storage Requirements 12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity Angiopoietin-like Protein 3/ANGPTL3
Species Mouse
Reconstitution Reconstitute at 400 μg/mL in water.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.