Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Angiopoietin-like 7 Protein

Catalog No. 4960AN025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaPO 4 and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in NaPO 4 and NaCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
4960AN025 Lyophilized from a 0.2 μm filtered solution in NaPO 4 and NaCl with BSA as a carrier protein.
4960AN025CF Lyophilized from a 0.2 μm filtered solution in NaPO 4 and NaCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 4960AN025 Supplier R&D Systems™ Supplier No. 4960AN025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Angiopoietin-like 7 Protein is derived from CHO. The Recombinant Mouse Angiopoietin-like 7 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in NaPO 4 and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 654812
Molecular Weight (g/mol) 37.9 kDa
Name Angiopoietin-like Protein 7/ANGPTL7
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Angiopoietin-like Protein 7/ANGPTL7 protein Met1-Pro337,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.