Learn More
R&D Systems™ Recombinant Mouse Angiopoietin-like Protein 2/ANGPTL2
Description
Specifications
Specifications
| Accession Number | AAD55358 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 23452 |
| Molecular Weight (g/mol) | 29 kDa |
| Name | angiopoietin-like 2, Angiopoietin-like protein 2, angiopoietin-related protein 2, ANGPTL2, ANGRP2, ARP2HARP, MGC8889 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Angiopoietin-like protein, (N-terminus) 2/ANGPTL2 protein, (N-terminus) Mouse ANGPTL2 (Ala260-His493) Accession &Num; AAD55358 HP GGGSGGGSGGGS HHHHHH (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.