Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse B7-H2 Fc Chimera Protein

Catalog No. 158B7100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
158B7100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 158B7100 Supplier R&D Systems™ Supplier No. 158B7100
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New rm B7-H2 is Available! Active on Mouse Th2 cells (D10 G4.1 cells) to induce IL-4 and has a His-tag!

Measured in a competitive binding assay. When rmICOS/Fc Chimera (Catalog # 168-CS) is immobilized at 1 μg/mL (100 μL/well), rmB7-H2 inhibits 50% binding of biotinylated rhB7-H2/Fc Chimera (2.5 ng/mL) at the concentration range of 80-120 ng/mL.

Specifications

Accession Number Q9JHJ8
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 23308
Molecular Weight (g/mol) M.W. (Observed): 80-90 kDa, reducing conditions; M.W. (theoretical): 53 kDa (monomer)
Name B7-H2 Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived mouse B7-H2 protein Mouse B7-H2 (Glu47 - Lys279) (N-terminus) Accession # Q9JHJ8 DIEGRMD Human IgG1 (Pro100 - Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Icosl
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein B7-H2/ICOSLG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.