Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse BAFF/BLyS/TNFSF13B Protein
SDP

Catalog No. 8876BF010CF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-76B-F010CF 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 88-76B-F010CF Supplier R&D Systems™ Supplier No. 8876BF010/CF
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse BAFF/BLyS/TNFSF13B Protein is derived from NS0. The Recombinant Mouse BAFF/BLyS/TNFSF13B Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9WU72
For Use With (Application) Bioactivity
Gene ID (Entrez) 10673
Molecular Weight (g/mol) Observed M.W.: 19-23kDa, reducing conditions, Theoretical M.W.: 21kDa
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse BAFF/BLyS/TNFSF13B protein Ala127-Leu309
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Common Name BAFF/BLyS/TNFSF13B
Gene Symbol Tnfsf13b
Biological Activity Measured in a cell proliferation assay using anti-IgM stimulated mouse B cells. The ED50 for this effect is typically 0.1-0.5ng/mL in the presence of goat anti-mouse IgMμ chain.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Ala127-Leu309
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.