Learn More
R&D Systems™ Recombinant Mouse BAFFR/TNFRSF13C Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 115650 |
| Molecular Weight (g/mol) | 33 kDa (monomer) |
| Name | BAFFR/TNFRSF13C |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse BAFF R/TNFRSF13C protein, (N-terminus) Mouse BAFF R (Ser10-Ala 71) Accession &Num; Q9D8D0 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.