Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES and KCl. |
| Gene ID (Entrez) | 12048 |
| Molecular Weight (g/mol) | 25 kDa |
| Name | Bcl-xL Minus C-Terminus |
| Quantity | 50 μg |
| Source | E. coli-derived mouse Bcl-xL protein Ser2-Arg212,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Monomer and disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
