Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse BID (Caspase-8-cleaved) Protein

Catalog No. 883M8050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
883M8050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 883M8050 Supplier R&D Systems™ Supplier No. 883M8050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce cytochrome c release from isolated mouse liver mitochondria. The typical EC50 for cytochrome c releasing activity is <100 nM. Cytochrome c is quantified using the Rat/Mouse Cytochrome c Quantikine™ ELISA Kit (Catalog # MCTC0 ). The EC50 for the desired application should be determined. Uncleaved Recombinant Mouse BID is available (Catalog # 860-MB ).

Specifications

Accession Number P70444
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES-KOH and KCl.
Gene ID (Entrez) 637
Molecular Weight (g/mol) M.W. (Observed): 7 kDa and 15 kDa, under reducing and non-reducing conditions; M.W. (theoretical): 7 kDa (N-terminal fragment) & 15 kDa (C-terminal fragment)
Name BID Caspase-8-cleaved
Quantity 50 μg
Source E. coli-derived mouse BID protein Met1-Asp195
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Gene Symbol Bid
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein BID
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.