Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse BMP-5 Protein

Catalog No. 6176BM020
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
6176BM020 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
6176BM020CF Lyophilized from a 0.2μm filtered solution in HCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 6176BM020 Supplier R&D Systems™ Supplier No. 6176BM020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse BMP-5 Protein is derived from CHO. The Recombinant Mouse BMP-5 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gene ID (Entrez) 12160
Molecular Weight (g/mol) 15.6 kDa (monomer)
Name BMP-5
Quantity 20 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse BMP-5 protein Ala315-His452
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.