Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse BTNL9 Fc Chimera Protein
SDP

Catalog No. 9555BT050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9555BT050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9555BT050 Supplier R&D Systems™ Supplier No. 9555BT050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Learn More About B7-related Butryophilins as Immune Modulators View Webinar

Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.75-4.5 μg/mL.

Specifications

Accession Number Q8BJE2-1
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 153579
Molecular Weight (g/mol) M.W. (Observed): 60-73 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse BTNL9 protein Mouse BTNL9 (Asp36-Lys257) (N-terminus) Accession # Q8BJE2-1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias BTN3, butyrophilin 3, butyrophilin-like 9, butyrophilin-like protein 9, UNQ1900/PRO4346, VDLS1900
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BTNL9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.