Learn More
Description
Specifications
Specifications
| Accession Number | Q6PDJ1.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 57685 |
| Molecular Weight (g/mol) | M.W.-observed: 130-144 kDa, under reducing conditions., M.W.-theroretical: 145 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse CACHD1 protein Mouse CACHD1 (Asp52-Pro1109) (C-terminus) Accession # Q6PDJ1.1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | cache domain containing 1, Cache domain-containing protein 1, KIAA1573VWFA and cache domain-containing protein 1, RP4-655E10.1, von Willebrand factor type A and cache domain containing 1, VWCD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
