Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Carboxylesterase 31-like/EG13909

Catalog No. 6059CE010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6059CE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 6059CE010 Supplier R&D Systems™ Supplier No. 6059CE010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its esterase activity. The specific activity is >750 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q8VCU1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 13909
Molecular Weight (g/mol) M.W. (Observed): 62 kDa, reducing conditions; M.W. (theoretical): 60 kDa
Name Carboxylesterase 31-like/EG13909
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse Carboxylesterase 31-like/EG13909 protein Met1-Glu558, with a C-terminal 10-His tag Accession # Q8VCU1
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Monomer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Ces3b
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Carboxylesterase 31-like/EG13909
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.