Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Caspr2 His-tag Protein

Catalog No. 10196CR100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-196-CR100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-196-CR100 Supplier R&D Systems™ Supplier No. 10196CR100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contactin Associated Protein-Like 2 (CASPR2), also known as Cntnap2, is a neuronal type I transmembrane member of the neurexin family of adhesion molecules

  • CASPR2, along with CASPR1 (or Cntnap1), is required for radical and longitudinal organization of myelinated axons
  • CASPR2 is thought to be important for the control of neuron-glia interactions and is associated with alterations in synaptic structure, neuronal network activity, and an autism-related phenotype in mice

Specifications

Accession Number Q9CPW0
For Use With (Application) Bioactivity
Gene ID (Entrez) 26047
Molecular Weight (g/mol) 138 kDa
Purification Method >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived mouse Caspr2 protein Ala28-Ser1262, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.