Learn More
R&D Systems™ Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 20305 |
| Molecular Weight (g/mol) | 8.3 kDa |
| Name | CCL6/C10 Truncated, aa 42-116 |
| Quantity | 25 μg |
| Source | E. coli-derived mouse CCL6/C10 protein Gly42-Ala116 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.