Learn More
R&D Systems™ Recombinant Mouse CD177 Protein
Catalog No. 8186CD
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
81-86C-D 50 μg
1 options
This item is not returnable. View return policy
Catalog No. 81-86C-D Supplier R&D Systems™ Supplier No. 8186CD050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
The Recombinant Mouse CD177 Protein is derived from NS0. The Recombinant Mouse CD177 Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately at the recommended storage temperatures.
Specifications
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse CD177 protein Leu22-Ala786,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.