Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse CD27 Ligand/TNFSF7 Protein

Catalog No. 783CL050CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
783CL050CF Lyophilized from a 0.2 μm filtered solution in PBS.
783CL050 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 783CL050CF Supplier R&D Systems™ Supplier No. 783CL050/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to stimulate mouse T cell proliferation in the presence of anti-CD3. The ED50 for this effect is 1.25-5.0 μg/mL.

Specifications

Accession Number Q05A52
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 970
Molecular Weight (g/mol) M.W. (Observed): 30 kDa, reducing conditions; M.W. (theoretical): 19.2 kDa
Name CD27 Ligand/TNFSF7
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD27 Ligand/TNFSF7 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse CD27 Ligand (Gln47 - Pro195) (N-terminus) Accession # Q05A52 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd70
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein CD27 Ligand/TNFSF7/CD70
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.