Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse CD40/TNFRSF5 Fc Chimera Protein

Catalog No. 1215CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1215CD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1215CD050 Supplier R&D Systems™ Supplier No. 1215CD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Mouse CD40/TNFRSF5 Fc Chimera is immobilized at 1 μg/mL (100 μL/well), the concentration of Recombinant Mouse CD40 Ligand/TNFSF5 (Catalog # 8230-CL ) that produces 50% optimal binding is found to be 100-600 ng/mL.

Specifications

Accession Number P27512
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 958
Molecular Weight (g/mol) M.W. (Observed): 58-61 kDa, reducing conditions; M.W. (theoretical): 46 kDa (monomer)
Name CD40/TNFRSF5 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD40/TNFRSF5 protein Mouse CD40 (Val24-Arg193) (N-terminus) Accession # P27512 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd40
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD40/TNFRSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.