Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse CD42b/GPIb alpha Protein

Catalog No. 8428GP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8428GP050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8428GP050 Supplier R&D Systems™ Supplier No. 8428GP050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse CD42b/GPIb alpha Protein is derived from CHO. The Recombinant Mouse CD42b/GPIb alpha Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number O35930
For Use With (Application) Bioactivity
Gene ID (Entrez) 2811
Molecular Weight (g/mol) Observed M.W.: 108-150kDa, reducing conditions, Theoretical M.W.: 66kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse CD42b/GPIb alpha protein Gln17-Pro612,with a C-terminal 6-His tag Accession &Num; O35930
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Antigen CD42b-alpha, BP1BA, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP1BA, GPIb alpha, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Common Name CD42b/GPIb alpha
Gene Symbol Gp1ba
Biological Activity Measured by its binding ability in a functional ELISA. Recombinant Mouse CD42b/GPIb alpha is immobilized at 0.5μg/mL (100μL/well), the concentration of Recombinant Human vWF-A2 that produces 50% optimal binding response is approximately 0.1-0.5μg/mL in the presence of ristocetin.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Gln17-Pro612, with a C-terminal 6-His tag Accession # O35930
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.