Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse CD52 Fc Chimera Protein

Catalog No. 10712CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10712CD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10712CD050 Supplier R&D Systems™ Supplier No. 10712CD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human Siglec-10 Fc Chimera Protein (Catalog # 2130-SL ) is immobilized at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Mouse CD52 Fc Chimera (Catalog # 10712-CD) that produces 50% of the optimal binding response is found to be approximately 1.5-7.5 μg/mL.

Specifications

Accession Number Q64389.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1043
Molecular Weight (g/mol) M.W.-observed: 32-36 kDa & 38-42 kDa, under reducing conditions, M.W.-theroretical: 29 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD52 protein Mouse CD52 (Thr27-Ser47) (N-terminus) Accession # Q64389.1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171 mP, Human epididymis-specific protein 5
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD52
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.