Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse CLEC-2/CLEC1B Protein

Catalog No. 9117CL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9117CL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9117CL050 Supplier R&D Systems™ Supplier No. 9117CL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse CLEC-2/CLEC1B Protein is derived from NS0. The Recombinant Mouse CLEC-2/CLEC1B Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9JL99
For Use With (Application) Bioactivity
Gene ID (Entrez) 51266
Molecular Weight (g/mol) Observed M.W.: 24-39kDa, reducing conditions, Theoretical M.W.: 21kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse CLEC-2/CLEC1B protein Met53-Leu229,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias 1810061I13Rik, CLEC1B, CLEC2, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721
Common Name CLEC-2/CLEC-1B
Gene Symbol Clec1b
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Mouse Podoplanin Fc Chimera is coated at 0.5μg/mL (100μL/well), Recombinant Mouse CLEC-2/CLEC1B binds with a typical ED50 of 0.1-0.5μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Met53-Leu229, with an N-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.