Learn More
R&D Systems™ Recombinant Mouse Coagulation Factor Xa Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in MES, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 14058 |
| Molecular Weight (g/mol) | 29 kDa & 10 kDa |
| Name | Coagulation Factor X |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Coagulation Factor X protein Met1-Asn481,with a C-terminal 10-His tag. Accession &Num; O88947 Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Recombinant Mouse Coagulation Factor X is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.