Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse COMP Protein

Catalog No. 8958CP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-58C-P050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-58C-P050 Supplier R&D Systems™ Supplier No. 8958CP050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce adhesion of ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.04-0.6 μg/mL.

Specifications

Accession Number Q9R0G6
For Use With (Application) Bioactivity
Gene ID (Entrez) 1311
Molecular Weight (g/mol) Observed M.W.: 85-115kDa, reducing conditions, Theoretical M.W.: 81kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse COMP/Thrombospondin-5 protein Gln20-Val755,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias cartilage oligomeric matrix protein, cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia1, multiple), cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia1, multiple), EDM1, EPD1, MED, MEDMGC131819, MGC149768, PSACH, pseudoachondroplasia (epiphyseal dysplasia 1, multiple), THBS5, thrombospondin-5, Thrombospondin5, TSP5
Common Name COMP/Thrombospondin-5
Gene Symbol Comp
Biological Activity Measured by its ability to induce adhesion of ATDC5 mouse chondrogenic cells. The ED50 for this effect is typically 0.15-0.9μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Gln20-Val755, with a C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.