Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse CRTAM Fc Chimera Protein

Catalog No. 3687CR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3687CR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3687CR050 Supplier R&D Systems™ Supplier No. 3687CR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding activity in a functional ELISA. Galiberet, L. et al. (2005) J. Biol. Chem. 280:21955. Immobilized rmCRTAM/Fc Chimera at 2 μg/mL (100 μL/well) can bind rmIGSF4A with a linear range of 0.15-10 μg/mL.

Specifications

Accession Number Q149L7
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 56253
Molecular Weight (g/mol) M.W. (Observed): 90 kDa, reducing conditions; M.W. (theoretical): 56.1 kDa (monomer)
Name CRTAM Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CRTAM protein Mouse CRTAM (Ala17 - Ser284) (N-terminus) Accession #Q149L7 IEGRMD Human IgG1 (Pro100 - Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein CRTAM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.