Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Cystatin F Protein

Catalog No. 4557PI010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4557PI010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4557PI010 Supplier R&D Systems™ Supplier No. 4557PI010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Cystatin F Protein is derived from NS0. The Recombinant Mouse Cystatin F Protein has been validated for the following applications: Inhibition Activity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl and Brij-35.
Molecular Weight (g/mol) 15 kDa
Name Cystatin F
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse Cystatin F protein Ala19-Gln144,with a C-terminal 6-His tag Accession &Num; O89098
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.