Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse DC-SIGN/CD209 Protein

Catalog No. 8345DC050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-45D-C050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-45D-C050 Supplier R&D Systems™ Supplier No. 8345DC050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse DC-SIGN/CD209 Protein is derived from NS0. The Recombinant Mouse DC-SIGN/CD209 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q91ZX1
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 170786
Molecular Weight (g/mol) 20kDa
Name DC-SIGN/CD209
Preparation Method Reconstitute at 100μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 21-26kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse DC-SIGN/CD209 protein Val73-Lys238,with an N-terminal HA tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Tyr (HA tag)
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.