Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse DcTRAILR1/TNFRSF23 Protein

Catalog No. 8479DT025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-79D-T025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-79D-T025 Supplier R&D Systems™ Supplier No. 8479DT025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse DcTRAIL R1/TNFRSF23 Protein is derived from NS0. The Recombinant Mouse DcTRAIL R1/TNFRSF23 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9ER63
For Use With (Application) Bioactivity
Gene ID (Entrez) 8718
Molecular Weight (g/mol) Observed M.W.: 23-30kDa, reducing conditions, Theoretical M.W.: 15kDa
Name DcTRAILR1/TNFRSF23
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse DcTRAIL R1/TNFRSF23 protein Met30-Ser156,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name DcTRAIL R1/TNFRSF23
Gene Symbol Tnfrsf23
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Mouse DcTRAIL R1/TNFSF23 is immobilized at 0.1μg/mL (100μL/well), the concentration of Recombinant Mouse TRAIL/TNFSF10 that produces 50% optimal binding response is approximately 2-12ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Met30-Ser156, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.