Learn More
R&D Systems™ Recombinant Mouse EG-VEGF/PK1 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 246691 |
| Molecular Weight (g/mol) | 9.6 kDa |
| Name | EG-VEGF/PK1 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse EG-VEGF/PK1 protein Ala20-Phe105 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.