Learn More
R&D Systems™ Recombinant Mouse Endorepellin His-tag Protein
Description
Specifications
Specifications
| Accession Number | NP_032331.2 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 3339 |
| Molecular Weight (g/mol) | 75.8 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Endorepellin/Perlecan protein Glu3683-Ser4383,with a C-terminal 6-His tag |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.