Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse ENPP-6 Protein

Catalog No. 8990EN020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-90E-N020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-90E-N020 Supplier R&D Systems™ Supplier No. 8990EN020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave O-(4-Nitrophenylphosphoryl) choline. The specific activity is >3,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number AAH96376
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 133121
Molecular Weight (g/mol) M.W. (Observed): 50-64 kDa, reducing conditions; M.W. (theoretical): 47 kDa
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived mouse ENPP-6 protein His23-Ser419, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias B830047L21Rik, EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 6, ectonucleotide pyrophosphatase/phosphodiesterase family member 6, E-NPP 6, ENPP6, GPC-Cpde, MGC33971, NPP6, NPP-6
Common Name ENPP6
Gene Symbol Enpp6
Biological Activity Measured by its ability to cleave O-(4-Nitrophenylphosphoryl) choline. The specific activity is >3,000 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived His23-Ser419, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ENPP6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.