Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Ephrin-A1 Fc Chimera Biotinylated Protein

Catalog No. BT602025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT602025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT602025 Supplier R&D Systems™ Supplier No. BT602025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

For Research Use Only

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Mouse Ephrin-A1 Fc Chimera Biotinylated Protein is derived from NS0. The Recombinant Mouse Ephrin-A1 Fc Chimera Biotinylated Protein has been validated for the following applications: Binding Activity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P52793
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
Gene ID (Entrez) 1942
Molecular Weight (g/mol) 46.8 kDa (monomer)
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse Ephrin-A1 protein, (N-terminus) Mouse Ephrin-A1 (Asp19-Ser182) Accession &Num; P52793 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked homodimer
Biological Activity 0.078 to 5 ng/mL
Product Type Recombinant protein
Conjugate Biotin
Species Mouse
Recombinant Recombinant
Protein Tag Yes
Reconstitution Reconstitute at 100 μg/mL with sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.