Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Ephrin-A4 Fc Chimera Biotinylated Protein

Catalog No. BT569
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT569 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT569 Supplier R&D Systems™ Supplier No. BT569
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Ephrin-A4 Fc Chimera Biotinylated Protein is derived from NS0. The Recombinant Mouse Ephrin-A4 Fc Chimera Biotinylated Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 13639
Molecular Weight (g/mol) 43.6 kDa (monomer)
Name Ephrin-A4 Fc Chimera
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse Ephrin-A4 protein, (N-terminus) Human Ephrin-A4 (Leu26-Gly176) Accession &Num; Q3UQB5 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Biotin
Recombinant Recombinant
Reconstitution Reconstitute with sterile PBS at 200 μg/mL.
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.