Learn More
Eurofins Discoverx Recombinant Mouse EphrinB2-Fc, 200 µg. (92-1089)
Supplier:  Eurofins Discoverx 921089
Encompass
Murine Ephrin-B2 (EPHB2) Fc Chimera is a fusion of EPHB2 to the C-terminal 6X histidine-tagged Fc region of IgG1. EPHB2 is a transmembrane protein-tyrosine kinase and member of the ephrin family. It is implicated in developmental processes, particularly of the nervous system, as well as pattern formation and heart morphogenesis and angiogenesis through regulation of cell adhesion and migration. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. NC1690911
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.